2,3-Diamino-5-bromo-6-(trifluoromethyl)-4(3H)-pyrimidinone ≥95%

Supplier: Matrix Scientific

045808-500MG 045808-1G 045808-5G
101872-712EA 375.38 USD
101872-712 101872-714 101872-716
2,3-Diamino-5-bromo-6-(trifluoromethyl)-4(3H)-pyrimidinone ≥95%
2,3-Diamino-5-bromo-6-(trifluoromethyl)-4(3H)-pyrimidinone

Hazard: Irritant

Formula: C₅H₄BrF₃N₄O
Melting Pt: 226…228 °C
MDL Number: MFCD00214813
CAS Number: 95095-46-8

Order Now

Specification Test Results

Melting point 226-228°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR