2-Amino-4,6-dimethylnicotinic acid ≥95%

Supplier: Matrix Scientific

045807-500MG 045807-1G 045807-5G
101872-706EA 424.62 USD
101872-706 101872-708 101872-710
2-Amino-4,6-dimethylnicotinic acid ≥95%
2-Amino-4,6-dimethylnicotinic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
Melting Pt: 223°C
MDL Number: MFCD00138571
CAS Number: 106837-89-2

Order Now

Specification Test Results

Melting point 223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR