5-(1-Phenoxyethyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

045806-500MG 045806-1G 045806-5G
101872-700EA 375.38 USD
101872-700 101872-702 101872-704
5-(1-Phenoxyethyl)-1H-pyrazole ≥95%
5-(1-Phenoxyethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₁₂N₂O
MW: 188.23 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD00139123
CAS Number: 321998-96-3

Order Now

Specification Test Results

Melting point 109-111°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR