2-Amino-4,6-dimethylnicotinamide ≥95%
Supplier: Matrix Scientific
045803-500MG
045803-1G
045803-5G
101872-682EA
747.69
USD
101872-682
101872-684
101872-686
2-Amino-4,6-dimethylnicotinamide ≥95%
2-Amino-4,6-dimethylnicotinamide
Hazard: Irritant
Formula:
C₈H₁₁N₃O Melting Pt: 162-166°C |
MDL Number:
MFCD00138572 CAS Number: 7144-20-9 |
Specification Test Results
Melting point | 162-166°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...