2-Chloro-4-(4-methylphenyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

045795-500MG 045795-1G 045795-5G
101872-642EA 556.92 USD
101872-642 101872-644 101872-646
2-Chloro-4-(4-methylphenyl)nicotinonitrile ≥95%
2-Chloro-4-(4-methylphenyl)nicotinonitrile

Hazard: Irritant

Formula: C₁₃H₉ClN₂
Melting Pt: 167…169 °C
MDL Number: MFCD08056677
CAS Number: 886362-03-4

Order Now

Specification Test Results

Melting point 167-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR