2-(3-Iodophenoxy)pyrimidine ≥95%
Supplier: Matrix Scientific
045793-500MG
045793-1G
045793-5G
101872-630EA
181.54
USD
101872-630
101872-632
101872-634
2-(3-Iodophenoxy)pyrimidine ≥95%
2-(3-Iodophenoxy)pyrimidine
Hazard: Irritant
Formula:
C₁₀H₇IN₂O Melting Pt: 92…94 °C |
MDL Number:
MFCD08056676 CAS Number: 886362-02-3 |
Specification Test Results
Melting point | 92-94°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...