4-(4-Methylphenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%

Supplier: Matrix Scientific

045792-500MG 045792-1G 045792-5G
101872-624EA 375.38 USD
101872-624 101872-626 101872-628
4-(4-Methylphenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile ≥95%
4-(4-Methylphenyl)-2-oxo-1,2-dihydro-3-pyridinecarbonitrile

Hazard: Irritant

Formula: C₁₃H₁₀N₂O
MW: 210.24 g/mol
Melting Pt: 238…240 °C
MDL Number: MFCD08056675
CAS Number: 886362-01-2

Order Now

Specification Test Results

Melting point 238-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR