1-{3-Chloro-4-[(4-methyl-3-nitrobenzyl)oxy]phenyl}-1-ethanone ≥95%

Supplier: Matrix Scientific

045787-500MG 045787-1G 045787-5G
101872-596EA 375.38 USD
101872-596 101872-598 101872-600
1-{3-Chloro-4-[(4-methyl-3-nitrobenzyl)oxy]phenyl}-1-ethanone ≥95%
1-{3-Chloro-4-[(4-methyl-3-nitrobenzyl)oxy]phenyl}-1-ethanone

Hazard: Irritant

Formula: C₁₆H₁₄ClNO₄
Melting Pt: 134…135 °C
MDL Number: MFCD06659605
CAS Number: 885950-07-2

Order Now

Specification Test Results

Melting point 134-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR