imidazo[1,2-a]pyridine-6-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: Imidazo[1,2-a]pyridine-6-carbonitrile, 6-Cyanoimidazo[1,2-a]pyridine

045785-1G 045785-5G
101872-586EA 130.77 USD
101872-586 101872-588
imidazo[1,2-a]pyridine-6-carbonitrile ≥95%
imidazo[1,2-a]pyridine-6-carbonitrile

Hazard: Irritant

Formula: C₈H₅N₃
MW: 143.149 g/mol
Melting Pt: 165…167 °C
MDL Number: MFCD06659603
CAS Number: 106850-34-4

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR