4-Methyl-6-(3-thienyl)-2-pyrimidinamine ≥95%

Supplier: Matrix Scientific

045782-500MG 045782-1G 045782-5G
101872-566EA 375.38 USD
101872-566 101872-568 101872-570
4-Methyl-6-(3-thienyl)-2-pyrimidinamine ≥95%
4-Methyl-6-(3-thienyl)-2-pyrimidinamine

Hazard: Irritant

Formula: C₉H₉N₃S
Melting Pt: 200…203 °C
MDL Number: MFCD06659600
CAS Number: 885950-01-6

Order Now

Specification Test Results

Melting point 200-203°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR