Methyl 5-iodo-2-oxo-1,2-dihydropyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl-5-iodo-2-oxo-1,2-dihydro-3-pyridinecarboxylate

045777-500MG 045777-1G 045777-5G
101872-536EA 150.77 USD
101872-536 101872-538 101872-540
Methyl 5-iodo-2-oxo-1,2-dihydropyridine-3-carboxylate ≥95%
Methyl 5-iodo-2-oxo-1,2-dihydropyridine-3-carboxylate

Hazard: Irritant

Formula: C₇H₆INO₃
MW: 279.04 g/mol
Melting Pt: 199…201 °C
MDL Number: MFCD06659597
CAS Number: 116387-40-7

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR