Methyl-3-[2-amino-4-(trifluoromethyl)anilino]propanoate ≥95%

Supplier: Matrix Scientific

045774-500MG 045774-1G 045774-5G
101872-526EA 375.38 USD
101872-526 101872-528 101872-530
Methyl-3-[2-amino-4-(trifluoromethyl)anilino]propanoate ≥95%
Methyl-3-[2-amino-4-(trifluoromethyl)anilino]propanoate

Hazard: Irritant

Formula: C₁₁H₁₃F₃N₂O₂
Melting Pt: 60…62 °C
MDL Number: MFCD06659595
CAS Number: 885949-99-5

Order Now

Specification Test Results

Melting point 60-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR