4-[2-Nitro-4-(trifluoromethyl)anilino]butanoic acid ≥95%

Supplier: Matrix Scientific

045773-500MG 045773-1G 045773-5G
101872-520EA 278.46 USD
101872-520 101872-522 101872-524
4-[2-Nitro-4-(trifluoromethyl)anilino]butanoic acid ≥95%
4-[2-Nitro-4-(trifluoromethyl)anilino]butanoic acid

Hazard: Irritant

Formula: C₁₁H₁₁F₃N₂O₄
Melting Pt: 95…98 °C
MDL Number: MFCD00586806
CAS Number: 65783-35-9

Order Now

Specification Test Results

Melting point 95-98°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR