7-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Amino-4-methyl-2H-benzo[b][1,4]oxazin-3(4H)-one

045772-500MG 045772-1G 045772-5G
101872-514EA 150.77 USD
101872-514 101872-516 101872-518
7-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one ≥95%
7-Amino-4-methyl-2H-1,4-benzoxazin-3(4H)-one

Hazard: Irritant

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
Melting Pt: 141…143 °C
MDL Number: MFCD06659594
CAS Number: 141068-81-7

Order Now

Specification Test Results

Melting point 141-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR