tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(4-nitrobenzoyl)piperazine-1-carboxylate

045771-500MG 045771-1G 045771-5G
101872-508EA 278.46 USD
101872-508 101872-510 101872-512
tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-(4-nitrobenzoyl)tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₁N₃O₅
MW: 335.36 g/mol
Melting Pt: 144 °C
MDL Number: MFCD06659593
CAS Number: 509073-62-5

Order Now

Specification Test Results

Melting point 144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR