3-(Morpholinosulfonyl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

045769-500MG 045769-1G 045769-5G
101872-496EA 104.62 USD
101872-496 101872-498 101872-500
3-(Morpholinosulfonyl)benzenecarboxylic acid ≥95%
3-(Morpholinosulfonyl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃NO₅S
Melting Pt: 195 °C
MDL Number: MFCD00622729
CAS Number: 299181-75-2

Order Now

Specification Test Results

Melting point 195°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR