5-Chloro-2-methyl-6-nitro-1,3-benzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-methyl-6-nitrobenzo[d]oxazole

045766-500MG 045766-1G 045766-5G
101872-480EA 100 USD
101872-480 101872-482 101872-484
5-Chloro-2-methyl-6-nitro-1,3-benzoxazole ≥95%
5-Chloro-2-methyl-6-nitro-1,3-benzoxazole

Hazard: Irritant

Formula: C₈H₅ClN₂O₃
MW: 212.59 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD00804971
CAS Number: 13452-16-9

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR