5-Chloro-6-nitro-1,3-benzoxazole ≥95%
Supplier: Matrix Scientific
045765-500MG
045765-1G
045765-5G
101872-474EA
100
USD
101872-474
101872-476
101872-478
5-Chloro-6-nitro-1,3-benzoxazole ≥95%
5-Chloro-6-nitro-1,3-benzoxazole
Hazard: Irritant
Formula:
C₇H₃ClN₂O₃ Melting Pt: 88…89 °C |
MDL Number:
MFCD05663828 CAS Number: 116549-11-2 |
Specification Test Results
Melting point | 95-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...