8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

045761-500MG 045761-1G 045761-5G
101872-454EA 55.38 USD
101872-454 101872-456 101872-458
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine ≥95%
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₄ClF₃N₂
Melting Pt: 107…109 °C
MDL Number: MFCD06496224
CAS Number: 178488-36-3

Order Now

Specification Test Results

Melting point 107-109°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR