Ethyl-6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate ≥95%
Supplier: Matrix Scientific
045759-500MG
045759-1G
101872-446EA
375.38
USD
101872-446
101872-448
Ethyl-6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate ≥95%
Ethyl 6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₁H₈ClF₃N₂O₂ Melting Pt: 100…102 °C |
MDL Number:
MFCD06637317 CAS Number: 420130-58-1 |
Specification Test Results
Melting point | 100-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...