N,N-Dimethyl(6-methylimidazo[1,2-a]pyridin-3-yl)methanamine ≥95%
Supplier: Matrix Scientific
Synonyms:
3-[(Dimethylamino)methyl]-6-methylimidazo[1,2-a]pyridine
045757-500MG
045757-1G
045757-5G
101872-440EA
375.38
USD
101872-440
101872-442
101872-444
N,N-Dimethyl(6-methylimidazo[1,2-a]pyridin-3-yl)methanamine ≥95%
N,N-Dimethyl(6-methylimidazo[1,2-a]pyridin-3-yl)methanamine
Hazard: Irritant
Formula:
C₁₁H₁₅N₃ MW: 189.26 g/mol Melting Pt: 82…84 °C |
MDL Number:
MFCD06200990 CAS Number: 885949-46-2 |
Specification Test Results
Melting point | 82-84°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...