6-Nitroimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

045755-1G 045755-5G 045755-25G
101872-428EA 23.08 USD
101872-428 101872-430 101872-432
6-Nitroimidazo[1,2-a]pyridine ≥95%
6-Nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₅N₃O₂
MW: 163.1300 g/mol
Melting Pt: 229-231°C
MDL Number: MFCD05863333
CAS Number: 25045-82-3

Order Now

Specification Test Results

Melting point 229-231°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR