Ethyl-5-aminobenzo[b]thiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

045750-500MG 045750-1G 045750-5G
101872-416EA 150.77 USD
101872-416 101872-418 101872-420
Ethyl-5-aminobenzo[b]thiophene-2-carboxylate ≥95%
Ethyl 5-aminobenzo[b]thiophene-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₁NO₂S
Melting Pt: 92…95 °C
CAS Number: 25785-10-8

Order Now

Specification Test Results

Melting point 92-95°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR