2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulfanyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(2-Hydroxy-3,3,3-trifluoroprop-1-yl)thio]benzoic acid

045745-500MG 045745-1G
101872-402EA 921.54 USD
101872-402 101872-404
2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulfanyl]benzenecarboxylic acid ≥95%
2-[(3,3,3-Trifluoro-2-hydroxypropyl)sulfanyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₀H₉F₃O₃S
Melting Pt: 98…100 °C
MDL Number: MFCD05663825
CAS Number: 886361-21-3

Order Now

Specification Test Results

Melting point 98-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR