3-[2-(4-Methoxyphenoxy)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045743-500MG 045743-1G 045743-5G
101872-392EA 921.54 USD
101872-392 101872-394 101872-396
3-[2-(4-Methoxyphenoxy)phenyl]propanoic acid ≥95%
3-[2-(4-Methoxyphenoxy)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₄
Melting Pt: 88…90 °C
MDL Number: MFCD05663823
CAS Number: 886361-17-7

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR