4-(2-Carboxybenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

045742-500MG 045742-1G 045742-5G
101872-386EA 747.69 USD
101872-386 101872-388 101872-390
4-(2-Carboxybenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
4-(2-Carboxybenzoyl)-1-methyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₁NO₅
Melting Pt: 221…222 °C
MDL Number: MFCD05663822
CAS Number: 886361-15-5

Order Now

Specification Test Results

Melting point 221-222°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR