4-(4-Methoxybenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

045741-500MG 045741-1G
101872-382EA 747.69 USD
101872-382 101872-384
4-(4-Methoxybenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(4-Methoxybenzoyl)-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₁NO₄
Melting Pt: 210…214 °C
MDL Number: MFCD05663821
CAS Number: 886361-13-3

Order Now

Specification Test Results

Melting point 210-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR