4-(3-Methoxybenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

045739-500MG 045739-1G
101872-374EA 747.69 USD
101872-374 101872-376
4-(3-Methoxybenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(3-Methoxybenzoyl)-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₁NO₄
Melting Pt: 163…166 °C
MDL Number: MFCD05663819
CAS Number: 886361-11-1

Order Now

Specification Test Results

Melting point 163-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR