1-(3-Nitrobenzoyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

045737-500MG 045737-1G 045737-5G
101872-362EA 181.54 USD
101872-362 101872-364 101872-366
1-(3-Nitrobenzoyl)-4-piperidinecarboxylic acid ≥95%
1-(3-Nitrobenzoyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₅
MW: 278.27 g/mol
Melting Pt: 185…188 °C
MDL Number: MFCD02070165
CAS Number: 352673-01-9

Order Now

Specification Test Results

Melting point 185-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR