3-[2-(4-Methylphenoxy)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045736-500MG 045736-1G 045736-5G
101872-356EA 921.54 USD
101872-356 101872-358 101872-360
3-[2-(4-Methylphenoxy)phenyl]propanoic acid ≥95%
3-[2-(4-Methylphenoxy)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₃
Melting Pt: 86…88 °C
MDL Number: MFCD05663817
CAS Number: 886361-09-7

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR