3-[2-(4-Chlorophenoxy)phenyl]propanoic acid ≥95%

Supplier: Matrix Scientific

045734-500MG 045734-1G 045734-5G
101872-344EA 921.54 USD
101872-344 101872-346 101872-348
3-[2-(4-Chlorophenoxy)phenyl]propanoic acid ≥95%
3-[2-(4-Chlorophenoxy)phenyl]propanoic acid

Hazard: Irritant

Formula: C₁₅H₁₃ClO₃
Melting Pt: 87…89 °C
MDL Number: MFCD05663816
CAS Number: 886361-07-5

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR