4-Propionyl-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

045733-500MG 045733-1G 045733-5G
101872-338EA 438.46 USD
101872-338 101872-340 101872-342
4-Propionyl-1H-pyrrole-2-carboxylic acid ≥95%
4-Propionyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.17 g/mol
Melting Pt: 216…220 °C
MDL Number: MFCD06200986
CAS Number: 111468-94-1

Order Now

Specification Test Results

Melting point 216-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR