4-(2,4-Dichlorobenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

045732-500MG 045732-1G
101872-334EA 556.92 USD
101872-334 101872-336
4-(2,4-Dichlorobenzoyl)-1H-pyrrole-2-carboxylic acid ≥95%
4-(2,4-Dichlorobenzoyl)-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₇Cl₂NO₃
Melting Pt: 121…125 °C
MDL Number: MFCD05663815
CAS Number: 886361-05-3

Order Now

Specification Test Results

Melting point 121-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR