(E)-3-[2-(3,5-Difluorophenoxy)phenyl]-2-propenoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: trans-2-(3,5-Difluorophenoxy)cinnamic acid

045728-500MG 045728-1G 045728-5G
101872-328EA 747.69 USD
101872-328 101872-330 101872-332
(E)-3-[2-(3,5-Difluorophenoxy)phenyl]-2-propenoic acid ≥95%
(E)-3-[2-(3,5-Difluorophenoxy)phenyl]-2-propenoic acid

Hazard: Irritant

Formula: C₁₅H₁₀F₂O₃
MW: 276.24 g/mol
Melting Pt: 185…188 °C
MDL Number: MFCD05663813
CAS Number: 886361-03-1

Order Now

Specification Test Results

Melting point 185-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR