4-(3,5-Difluorophenoxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

045727-500MG 045727-1G 045727-5G
101872-322EA 204.62 USD
101872-322 101872-324 101872-326
4-(3,5-Difluorophenoxy)benzaldehyde ≥95%
4-(3,5-Difluorophenoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₃H₈F₂O₂
Melting Pt: 57-60°C
MDL Number: MFCD05663812
CAS Number: 886361-01-9

Order Now

Specification Test Results

Melting point 57-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR