3-(2-Methyl-1,3-dioxolan-2-yl)aniline ≥95%

Supplier: Matrix Scientific

045725-500MG 045725-1G 045725-5G
101872-310EA 375.38 USD
101872-310 101872-312 101872-314
3-(2-Methyl-1,3-dioxolan-2-yl)aniline ≥95%
3-(2-Methyl-1,3-dioxolan-2-yl)aniline

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
Melting Pt: 72…74 °C
MDL Number: MFCD00020904
CAS Number: 51226-14-3

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR