2,4,6-Trimethoxyaniline hydrochloride ≥95%

Supplier: Matrix Scientific

045724-500MG 045724-1G 045724-5G
101872-304EA 747.69 USD
101872-304 101872-306 101872-308
2,4,6-Trimethoxyaniline hydrochloride ≥95%
2,4,6-Trimethoxyaniline hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO₃
Melting Pt: 220-225°C
MDL Number: MFCD00211326
CAS Number: 102438-99-3

Order Now

Specification Test Results

Melting point 220-225°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR