1-(4-Bromophenyl)-2,5-dimethylpyrrole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Bromophenyl)-2,5-dimethyl-1H-pyrrole

045723-500MG 045723-1G 045723-5G
101872-298EA 181.54 USD
101872-298 101872-300 101872-302
1-(4-Bromophenyl)-2,5-dimethylpyrrole ≥95%
1-(4-Bromophenyl)-2,5-dimethylpyrrole

Hazard: Irritant

Formula: C₁₂H₁₂BrN
Melting Pt: 75…77 °C
MDL Number: MFCD00277042
CAS Number: 5044-24-6

Order Now

Specification Test Results

Melting point 75-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR