3-(tert-Butyl)-1-phenyl-1H-pyrazole-5-carboxylic acid ≥95%

Supplier: Matrix Scientific

045722-500MG 045722-1G 045722-5G
101872-292EA 124.62 USD
101872-292 101872-294 101872-296
3-(tert-Butyl)-1-phenyl-1H-pyrazole-5-carboxylic acid ≥95%
3-(tert-Butyl)-1-phenyl-1H-pyrazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂
Melting Pt: 204…206 °C
MDL Number: MFCD02646688
CAS Number: 93045-47-7

Order Now

Specification Test Results

Melting point 204-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR