Ethyl-1-[4-amino-2-(methoxycarbonyl)phenyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

045721-500MG 045721-1G 045721-5G
101872-286EA 556.92 USD
101872-286 101872-288 101872-290
Ethyl-1-[4-amino-2-(methoxycarbonyl)phenyl]-4-piperidinecarboxylate ≥95%
Ethyl-1-[4-amino-2-(methoxycarbonyl)phenyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₆H₂₂N₂O₄
Melting Pt: 58…60 °C
MDL Number: MFCD03791246
CAS Number: 886360-83-4

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR