1-(2-Carboxy-4-nitrophenyl)-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
045719-500MG
045719-1G
045719-5G
101872-274EA
556.92
USD
101872-274
101872-276
101872-278
1-(2-Carboxy-4-nitrophenyl)-4-piperidinecarboxylic acid ≥95%
1-(2-Carboxy-4-nitrophenyl)-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₄N₂O₆ Melting Pt: 222…225 °C |
MDL Number:
MFCD03844859 CAS Number: 886360-81-2 |
Specification Test Results
Melting point | 222-225°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...