6-(4-Benzylpiperazino)nicotinic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
6-(4-Benzylpiperazin-1-yl)pyridine-3-carboxylic acid
045716-500MG
045716-1G
045716-5G
101872-262EA
576.92
USD
101872-262
101872-264
101872-266
6-(4-Benzylpiperazino)nicotinic acid ≥95%
6-(4-Benzylpiperazino)nicotinic acid
Hazard: Irritant
Formula:
C₁₇H₁₉N₃O₂ Melting Pt: 228…229 °C |
MDL Number:
MFCD03791243 CAS Number: 886360-80-1 |
Specification Test Results
Melting point | 228-229°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...