Ethyl-2-(methylamino)-4-oxo-4,5-dihydro-3-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

045714-500MG 045714-1G 045714-5G
101872-250EA 278.46 USD
101872-250 101872-252 101872-254
Ethyl-2-(methylamino)-4-oxo-4,5-dihydro-3-thiophenecarboxylate ≥95%
Ethyl-2-(methylamino)-4-oxo-4,5-dihydro-3-thiophenecarboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₃S
Melting Pt: 163…166 °C
MDL Number: MFCD03791241
CAS Number: 886360-78-7

Order Now

Specification Test Results

Melting point 163-166°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR