Methyl-4'-cyano[1,1'-biphenyl]-4-carboxylate ≥95%

Supplier: Matrix Scientific

045713-500MG 045713-1G 045713-5G
101872-244EA 556.92 USD
101872-244 101872-246 101872-248
Methyl-4'-cyano[1,1'-biphenyl]-4-carboxylate ≥95%
Methyl-4'-cyano[1,1'-biphenyl]-4-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₁NO₂
Melting Pt: 130…133 °C
MDL Number: MFCD03791240
CAS Number: 89900-95-8

Order Now

Specification Test Results

Melting point 130-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR