3-(4-Chlorophenyl)-2-(phenylsulfonyl)acrylonitrile ≥95%

Supplier: Matrix Scientific

045712-500MG 045712-1G 045712-5G
101872-238EA 269.23 USD
101872-238 101872-240 101872-242
3-(4-Chlorophenyl)-2-(phenylsulfonyl)acrylonitrile ≥95%
3-(4-Chlorophenyl)-2-(phenylsulfonyl)acrylonitrile

Hazard: Irritant

Formula: C₁₅H₁₀ClNO₂S
Melting Pt: 152…154 °C
MDL Number: MFCD03425821
CAS Number: 50709-88-1

Order Now

Specification Test Results

Melting point 152-154°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR