2-Amino-N-(2-propynyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-N-(prop-2-yn-1-yl)benzamide

045710-500MG 045710-1G 045710-5G
101872-226EA 747.69 USD
101872-226 101872-228 101872-230
2-Amino-N-(2-propynyl)benzenecarboxamide ≥95%
2-Amino-N-(2-propynyl)benzenecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
Melting Pt: 100…101 °C
MDL Number: MFCD03425820
CAS Number: 4943-83-3

Order Now

Specification Test Results

Melting point 100-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR