Methyl-2-hydroxy-4-(1H-pyrrol-1-yl)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-hydroxy-4-(1H-pyrrol-1-yl)benzoate

045709-500MG 045709-1G 045709-5G
101872-220EA 315.38 USD
101872-220 101872-222 101872-224
Methyl-2-hydroxy-4-(1H-pyrrol-1-yl)benzenecarboxylate ≥95%
Methyl-2-hydroxy-4-(1H-pyrrol-1-yl)benzenecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MW: 217.23 g/mol
Melting Pt: 132…135 °C
MDL Number: MFCD02083001
CAS Number: 454473-64-4

Order Now

Specification Test Results

Melting point 132-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR