Ethyl-1-(2-cyanophenyl)-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

045708-500MG 045708-1G 045708-5G
101872-214EA 67.69 USD
101872-214 101872-216 101872-218
Ethyl-1-(2-cyanophenyl)-4-piperidinecarboxylate ≥95%
Ethyl-1-(2-cyanophenyl)-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₂
MW: 258.32 g/mol
Melting Pt: 45…47 °C
MDL Number: MFCD04117820
CAS Number: 357670-16-7

Order Now

Specification Test Results

Melting point 45-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR