Ethyl-5-(aminocarbonyl)-4-methyl-6-thioxo-1,6-dihydro-3-pyridinecarboxylate ≥95%

Supplier: Matrix Scientific

045706-500MG 045706-1G
101872-204EA 375.38 USD
101872-204 101872-206
Ethyl-5-(aminocarbonyl)-4-methyl-6-thioxo-1,6-dihydro-3-pyridinecarboxylate ≥95%
Ethyl-5-(aminocarbonyl)-4-methyl-6-thioxo-1,6-dihydro-3-pyridinecarboxylate

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₃S
MW: 240.28 g/mol
Melting Pt: 271…273 °C
MDL Number: MFCD03425818
CAS Number: 171113-26-1

Order Now

Specification Test Results

Melting point 271-273°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR