4-(1H-1,2,3-Benzotriazol-1-ylmethyl)phenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[(1H-1,2,3-Benzotriazol-1-yl)methyl]aniline

045704-500MG 045704-1G 045704-5G
101872-192EA 556.92 USD
101872-192 101872-194 101872-196
4-(1H-1,2,3-Benzotriazol-1-ylmethyl)phenylamine ≥95%
4-(1H-1,2,3-Benzotriazol-1-ylmethyl)phenylamine

Hazard: Irritant

Formula: C₁₃H₁₂N₄
MW: 224.27 g/mol
Melting Pt: 140…143 °C
MDL Number: MFCD00960990
CAS Number: 129075-89-4

Order Now

Specification Test Results

Melting point 140-143°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR